Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Iodobenzoyl chloride, 98%

Click to view available options
Quantity:
5g
25g
100g
1711-02-0
C7H4ClIO
266.462
MFCD00001058
NJAKCIUOTIPYED-UHFFFAOYSA-N
p-iodobenzoyl chloride,benzoyl chloride, 4-iodo,benzoyl chloride, p-iodo,4-iodobenzoic acid chloride,4-iodobenzoylchloride,4-iodo-benzoyl chloride,4iodobenzoyl chloride,4-iodo-benzoylchlorid,rho-iodobenzoyl chloride,4-iodo benzoyl chloride
74373
4-iodobenzoyl chloride
C1=CC(=CC=C1C(=O)Cl)I
1711-02-0
C7H4ClIO
266.462
MFCD00001058
NJAKCIUOTIPYED-UHFFFAOYSA-N
p-iodobenzoyl chloride,benzoyl chloride, 4-iodo,benzoyl chloride, p-iodo,4-iodobenzoic acid chloride,4-iodobenzoylchloride,4-iodo-benzoyl chloride,4iodobenzoyl chloride,4-iodo-benzoylchlorid,rho-iodobenzoyl chloride,4-iodo benzoyl chloride
74373
4-iodobenzoyl chloride
C1=CC(=CC=C1C(=O)Cl)I
Applications
4-Iodobenzoyl chloride is employed in the preparation of pyrroles by reaction with imines and acetylenes mediated by isocyanides using palladium catalysis. It is also used in the preparation of N-(1-benzylpyrrolidin-3-yl)arylbenzamides, potential human dopamine D4 antagonists. Further, it is used to prepare [2]rotaxane monomer and poly[2] rotaxane. In addition to this, it is important in the modification of poly(allylamine) in order to make the polymer X-ray visible.

Solubility
Soluble in toluene and benzene.

Notes
Moisture sensitive. Incompatible with strong bases and water.
Quantity 25g
Melting Point 61°C to 65°C
Flash Point >110°C (230°F)
Boiling Point 120°C to 121°C (1mmHg)
UN Number UN3261
Beilstein 774718
Formula Weight 266.47
Percent Purity 98%
Chemical Name or Material 4-Iodobenzoyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA