Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Iodobenzyl bromide, 97%

Click to view available options
Quantity:
5g
25g
100g
16004-15-2
C7H6BrI
296.933
MFCD00209656
BQTRMYJYYNQQGK-UHFFFAOYSA-N
4-iodobenzyl bromide,1-bromomethyl-4-iodobenzene,4-iodobenzylbromide,p-iodobenzyl bromide,1-bromomethyl-4-iodo-benzene,alpha-bromo-4-iodotoluene,4-bromomethyl-1-iodobenzene,benzene, 1-bromomethyl-4-iodo,4-iodo benzyl bromide
954258
1-(bromomethyl)-4-iodobenzene
C1=CC(=CC=C1CBr)I
16004-15-2
C7H6BrI
296.933
MFCD00209656
BQTRMYJYYNQQGK-UHFFFAOYSA-N
4-iodobenzyl bromide,1-bromomethyl-4-iodobenzene,4-iodobenzylbromide,p-iodobenzyl bromide,1-bromomethyl-4-iodo-benzene,alpha-bromo-4-iodotoluene,4-bromomethyl-1-iodobenzene,benzene, 1-bromomethyl-4-iodo,4-iodo benzyl bromide
954258
1-(bromomethyl)-4-iodobenzene
C1=CC(=CC=C1CBr)I
Applications
4-Iodobenzyl bromide is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents, bases and amines.
Sensitivity Light sensitive
Quantity 100g
Boiling Point 140°C (15mmHg)
Odor Irritating
Melting Point 78°C to 82°C
UN Number UN3261
Beilstein 2325160
Solubility Information Insoluble in water.
Formula Weight 296.94
Percent Purity 97%
Chemical Name or Material 4-Iodobenzyl bromide
Show More Show Less

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures