Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodobenzyl bromide, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
16004-15-2
C7H6BrI
296.93
BQTRMYJYYNQQGK-UHFFFAOYSA-N
4-iodobenzyl bromide,1-bromomethyl-4-iodobenzene,4-iodobenzylbromide,p-iodobenzyl bromide,1-bromomethyl-4-iodo-benzene,alpha-bromo-4-iodotoluene,4-bromomethyl-1-iodobenzene,benzene, 1-bromomethyl-4-iodo,4-iodo benzyl bromide
954258
1-(bromomethyl)-4-iodobenzene
C1=CC(=CC=C1CBr)I
16004-15-2
C7H6BrI
296.93
BQTRMYJYYNQQGK-UHFFFAOYSA-N
4-iodobenzyl bromide,1-bromomethyl-4-iodobenzene,4-iodobenzylbromide,p-iodobenzyl bromide,1-bromomethyl-4-iodo-benzene,alpha-bromo-4-iodotoluene,4-bromomethyl-1-iodobenzene,benzene, 1-bromomethyl-4-iodo,4-iodo benzyl bromide
954258
1-(bromomethyl)-4-iodobenzene
C1=CC(=CC=C1CBr)I
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
Boiling Point 140.0°C (15.0 mmHg)
Melting Point 77.0°C to 82.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula IC6H4CH2Br
Formula Weight 296.93
Physical Form Adhering Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Iodobenzyl bromide
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger