Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodobiphenyl 97.0+%, TCI America™
SDP

Catalog No. I078525G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1591-31-7
Molecular Formula C12H9I
Molecular Weight (g/mol) 280.11
MDL Number MFCD00019028
InChI Key NXYICUMSYKIABQ-UHFFFAOYSA-N
Synonym 4-iodobiphenyl, 4-iodo-1,1'-biphenyl, 1,1'-biphenyl, 4-iodo, p-iodobiphenyl, 4-iododiphenyl, p-phenyliodobenzene, 4-biphenyl iodide, biphenyl, 4-iodo, 1-iodo-4-phenyl-benzene, 4-iodo-1-phenylbenzene
PubChem CID 15322
IUPAC Name 4-iodo-1,1'-biphenyl
SMILES IC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 112°C
Color White-Yellow
Boiling Point 320°C
Quantity 25 g
UN Number 3077
Formula Weight 280.11
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodobiphenyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.