Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodoimidazole, 95%, Acros Organics™

Click to view available options
Quantity:
1g
5g
71759-89-2
C3H3IN2
193.98
BHCMXJKPZOPRNN-UHFFFAOYSA-N
4-iodoimidazole,4-iodo-1h-imidazole,4 5-iodoimidazole,4 5-iodo-1 h-imidazole,4-iodo-1 h-imidazole,4-iodo-3h-imidazole,1h-imidazole, 4-iodo,5-iodoimidazole,4-iodo-imidazole,sftheqvilimkp@
606522
5-iodo-1H-imidazole
C1=C(NC=N1)I
71759-89-2
C3H3IN2
193.98
BHCMXJKPZOPRNN-UHFFFAOYSA-N
4-iodoimidazole,4-iodo-1h-imidazole,4 5-iodoimidazole,4 5-iodo-1 h-imidazole,4-iodo-1 h-imidazole,4-iodo-3h-imidazole,1h-imidazole, 4-iodo,5-iodoimidazole,4-iodo-imidazole,sftheqvilimkp@
606522
5-iodo-1H-imidazole
C1=C(NC=N1)I
Packaging Glass Bottle
Quantity 1g
Infrared Spectrum Authentic
Melting Point 135.0°C to 136.0°C
Assay Percent Range 94% min. (HPLC)
Formula Weight 193.98
Percent Purity 95%
Chemical Name or Material 4-Iodoimidazole

GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning