Learn More
4-Iodophenol, 98+%, Thermo Scientific Chemicals
Description
4-Iodophenol is a chemical used in chemiluminescence imaging assays and experiments, as diagnostic agents in order to detect cancer cells in patients. Also it is used in the synthesis of agonists towards the estrogen β receptor. Also used in wide range of medicals industrial applications as well as in human and animal nutrition products such as pharmaceutical intermediates, and polarizing films for Liquid Crystal Display (LCD) chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Iodophenol is a chemical used in chemiluminescence imaging assays and experiments, as diagnostic agents in order to detect cancer cells in patients. Also it is used in the synthesis of agonists towards the estrogen β receptor. Also used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display (LCD) chemicals.
Solubility
Soluble in alcohol and ether. Slightly soluble in water.
Notes
Light Sensitive, store in dark. Store in cool, dry place in tightly closed container. Store away from acid chlorides and acid anhydrides.
Specifications
Melting Point | 91°C to 95°C |
Boiling Point | 138°C (5 mmHg) |
Quantity | 5 g |
Beilstein | 1904544 |
Sensitivity | Light sensitive |
Merck Index | 14,5036 |
Solubility Information | Soluble in alcohol and ether. Slightly soluble in water. |
Formula Weight | 220.01 |
Percent Purity | ≥98% |
Chemical Name or Material | 4-Iodophenol |
Safety and Handling
Hazard Category | H302+H312+H332-H314-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: III |
EINECSNumber | 208-745-5 |
RTECSNumber | SL5600000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.