Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodophenol, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
500g
Packaging:
Glass bottle
540-38-5
C6H5IO
220.01
VSMDINRNYYEDRN-UHFFFAOYSA-N
p-iodophenol,phenol, 4-iodo,phenol, p-iodo,4-hydroxyiodobenzene,p-hydroxyiodobenzene,4-hydroxyphenyl iodide,4-iodo-phenol,unii-bh194bak0b,ccris 668,bh194bak0b
10894
CHEBI:43521
4-iodophenol
C1=CC(=CC=C1O)I
540-38-5
C6H5IO
220.01
VSMDINRNYYEDRN-UHFFFAOYSA-N
p-iodophenol,phenol, 4-iodo,phenol, p-iodo,4-hydroxyiodobenzene,p-hydroxyiodobenzene,4-hydroxyphenyl iodide,4-iodo-phenol,unii-bh194bak0b,ccris 668,bh194bak0b
10894
CHEBI:43521
4-iodophenol
C1=CC(=CC=C1O)I
Melting Point 89.0°C to 94.0°C
Packaging Glass bottle
Color Pink or Beige to Brown
Infrared Spectrum Authentic
Quantity 100g
Boiling Point 287.0°C
Assay Percent Range 98.5% min. (GC)
Linear Formula IC6H4OH
Beilstein 06,208
Merck Index 15,5079
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol and ether
Formula Weight 220.01
Physical Form Crystals or Crystalline Powder
Percent Purity 99%
Chemical Name or Material 4-Iodophenol
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Danger

EINECSNumber : 208-745-5