Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodophenoxyacetic acid, 90%, ACROS Organics™

Click to view available options
Quantity:
5g
1878-94-0
C8H7IO3
278.045
MFCD00014359
RKORKXFKXYYHAQ-UHFFFAOYSA-N
4-iodophenoxyacetic acid,2-4-iodophenoxy acetic acid,4-iodophenoxy acetic acid,p-iodophenoxyacetic acid,p-iodophenoxy acetic acid,acetic acid, 4-iodophenoxy,4-iodophenoxyacetic,4-iodophenoxyaceticacide,acmc-209eqm,4-iodo phenoxyacetic acid
74658
2-(4-iodophenoxy)acetic acid
C1=CC(=CC=C1OCC(=O)O)I
1878-94-0
C8H7IO3
278.045
MFCD00014359
RKORKXFKXYYHAQ-UHFFFAOYSA-N
4-iodophenoxyacetic acid,2-4-iodophenoxy acetic acid,4-iodophenoxy acetic acid,p-iodophenoxyacetic acid,p-iodophenoxy acetic acid,acetic acid, 4-iodophenoxy,4-iodophenoxyacetic,4-iodophenoxyaceticacide,acmc-209eqm,4-iodo phenoxyacetic acid
74658
2-(4-iodophenoxy)acetic acid
C1=CC(=CC=C1OCC(=O)O)I
Quantity 5g
Color White
Infrared Spectrum Authentic
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Linear Formula IC6H4OCHCOOH
Formula Weight 278.05
Physical Form Powder
Percent Purity ≥88%
Chemical Name or Material 4-Iodophenoxyacetic acid, 90%
Show More Show Less

EINECSNumber : 217-531-0