Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodophenylacetic acid, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
1798-06-7
C8H7IO2
262.05
FJSHTWVDFAUNCO-UHFFFAOYSA-N
4-iodophenylacetic acid,2-4-iodophenyl acetic acid,4-iodophenyl acetic acid,4-iodophenylaceticacid,p-iodophenylacetic acid,benzeneacetic acid, 4-iodo,4-iodo-phenyl-acetic acid,pubchem18835,4-iodoohenylacetic acid,4-iodobenzeneacetic acid
137214
2-(4-iodophenyl)acetic acid
C1=CC(=CC=C1CC(=O)O)I
1798-06-7
C8H7IO2
262.05
FJSHTWVDFAUNCO-UHFFFAOYSA-N
4-iodophenylacetic acid,2-4-iodophenyl acetic acid,4-iodophenyl acetic acid,4-iodophenylaceticacid,p-iodophenylacetic acid,benzeneacetic acid, 4-iodo,4-iodo-phenyl-acetic acid,pubchem18835,4-iodoohenylacetic acid,4-iodobenzeneacetic acid
137214
2-(4-iodophenyl)acetic acid
C1=CC(=CC=C1CC(=O)O)I
Melting Point 140°C to 141.5°C
Quantity 5g
Assay Percent Range 96% min. (HPLC)
Formula Weight 262.05
Percent Purity 97%
Chemical Name or Material 4-Iodophenylacetic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning