Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodophenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
5122-99-6
C6H6BIO2
247.82
PELJYVULHLKXFF-UHFFFAOYSA-N
4-iodophenyl boronic acid,4-iodobenzeneboronic acid,p-iodophenylboronic acid,4-iodophenylboronicacid,p-iodobenzeneboronic acid,p-iodophenyl boronic acid,boronic acid, 4-iodophenyl,4-boronoiodobenzene,pubchem6160,pubchem11574
151254
(4-iodophenyl)boronic acid
B(C1=CC=C(C=C1)I)(O)O
5122-99-6
C6H6BIO2
247.82
PELJYVULHLKXFF-UHFFFAOYSA-N
4-iodophenyl boronic acid,4-iodobenzeneboronic acid,p-iodophenylboronic acid,4-iodophenylboronicacid,p-iodobenzeneboronic acid,p-iodophenyl boronic acid,boronic acid, 4-iodophenyl,4-boronoiodobenzene,pubchem6160,pubchem11574
151254
(4-iodophenyl)boronic acid
B(C1=CC=C(C=C1)I)(O)O
Quantity 1g
Color Beige
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 322.0°C to 326.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: 25g/L
Formula Weight 247.82
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-Iodophenylboronic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning