Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (4-Iodophenylethynyl)trimethylsilane, 97%

Manufacturer:  Alfa Aesar™ H5525606

Quantity 5g

 View more versions of this product

Catalog No. AAH5525606

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Light sensitive.
Description & Specifications

Chemical Identifiers

134856-58-9
300.21
MFCD04974026
(4-Iodophenylethynyl)trimethylsilane

Specifications

134856-58-9
300.21
Light Sensitive
66° to 70°C
(4-Iodophenylethynyl)trimethylsilane
Powder
MFCD04974026
White to yellow
5g
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit