Alfa Aesar™ (4-Iodophenylethynyl)trimethylsilane, 97%

Manufacturer: Alfa Aesar™ H5525603

Quantity 1g

 View more versions of this product

Catalog No. AAH5525603

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 134856-58-9
    Formula Weight 300.21
    MDL Number MFCD04974026
    Chemical Name or Material (4-Iodophenylethynyl)trimethylsilane

    Specifications

    Formula Weight 300.21
    Melting Point 66° to 70°C
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Physical Form Powder
    Color White to yellow
    Quantity 1g
    Health Hazard 1 Warning
    MDL Number MFCD04974026
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    Sensitivity Light Sensitive
    CAS 134856-58-9
    Chemical Name or Material (4-Iodophenylethynyl)trimethylsilane
    Assay 97%

    Description

    Applications
    It is used as a pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Store at room temperature. Light sensitive.

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit