Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodotoluene 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
100g
Packaging:
Glass bottle
624-31-7
C7H7I
218.037
MFCD00001059
UDHAWRUAECEBHC-UHFFFAOYSA-N
4-iodotoluene,p-iodotoluene,benzene, 1-iodo-4-methyl,toluene, p-iodo,p-tolyl iodide,p-methyliodobenzene,1-methyl-4-iodobenzene,1-iodo-4-methyl-benzene,unii-g75kcn1am8,iodotoluene 4-
12207
1-iodo-4-methylbenzene
CC1=CC=C(C=C1)I
624-31-7
C7H7I
218.037
MFCD00001059
UDHAWRUAECEBHC-UHFFFAOYSA-N
4-iodotoluene,p-iodotoluene,benzene, 1-iodo-4-methyl,toluene, p-iodo,p-tolyl iodide,p-methyliodobenzene,1-methyl-4-iodobenzene,1-iodo-4-methyl-benzene,unii-g75kcn1am8,iodotoluene 4-
12207
1-iodo-4-methylbenzene
CC1=CC=C(C=C1)I
Color White to Yellow
Infrared Spectrum Authentic
Quantity 10g
Packaging Glass bottle
Melting Point 33°C to 36°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula IC6H4CH3
Beilstein 05, 312
Formula Weight 218.04
Physical Form Crystalline Low Melting Mass
Percent Purity ≥97.5%
Chemical Name or Material 4-Iodotoluene, 98%
Show More Show Less

EINECSNumber : 210-841-7

TSCA : TSCA