Alfa Aesar™ 4'-Isobutylacetophenone, 97%

Manufacturer: Alfa Aesar™ A1062418

Quantity 50g

 View more versions of this product

Catalog No. AAA1062418

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 38861-78-8
    Molecular Formula C12H16O
    Formula Weight 176.27
    MDL Number MFCD00027393
    UN Number UN1224
    Beilstein 1935275
    Chemical Name or Material 4'-Isobutylacetophenone

    Specifications

    Formula Weight 176.27
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    CAS 38861-78-8
    Quantity 50g
    Molecular Formula C12H16O
    MDL Number MFCD00027393
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    UN Number UN1224
    Beilstein 1935275
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
    Flash Point 54°C (129°F)
    Chemical Name or Material 4'-Isobutylacetophenone
    Assay 97%

    Description

    Applications
    4 ft.-isobutylacetophenone is an important starting material employed in the production of pharmaceuticals such as ibuprofen. It is also utilized in proteomics research. It is used as an intermediate in organic synthesis.

    Solubility
    Miscible with chloroform and methanol. Slightly miscible with water.

    Notes
    Keep the container tightly closed in a dry and well-ventilated place. Keep away from sources of ignition. Incompatible with reducing agents, strong oxidizing agents and strong bases.

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit