Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isocyanato-1-methyl-1H-pyrazole, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
1174064-57-3
C5H5N3O
123.12
MFCD14706124
TXMLNGHIFGKEFH-UHFFFAOYSA-N
4-isocyanato-1-methyl-1h-pyrazole
45790024
4-isocyanato-1-methylpyrazole
CN1C=C(C=N1)N=C=O
1174064-57-3
C5H5N3O
123.12
MFCD14706124
TXMLNGHIFGKEFH-UHFFFAOYSA-N
4-isocyanato-1-methyl-1h-pyrazole
45790024
4-isocyanato-1-methylpyrazole
CN1C=C(C=N1)N=C=O
Color Yellow
Boiling Point 203.0°C to 205.0°C
Quantity 5g
Flash Point 71°C
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 123.12
Physical Form Liquid, May Contain Fine Suspension
Percent Purity 95%
Chemical Name or Material 4-Isocyanato-1-methyl-1H-pyrazole
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritatio

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
IF ON SKI

GHS Signal Word: Danger