Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropoxycarbonylphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
342002-82-8
C10H13BO4
208.02
MFCD02259385
NIQHFKWRHJZMQU-UHFFFAOYSA-N
4-isopropoxycarbonylphenylboronic acid,4-isopropoxycarbonyl phenyl boronic acid,4-isopropoxycarbonyl benzeneboronic acid,propan-2-yl 4-dihydroxyboranyl benzoate,isopropyl 4-boronobenzoate,4-isopropoxycarbonyl phenylboronic acid,p-isopropoxycarbonyl phenylboronic acid,4-isopropoxycarbonylphenylboronicacid,4-propan-2-yloxycarbonylphenyl boronic acid,acmc-209i5o
2734703
(4-propan-2-yloxycarbonylphenyl)boronic acid
B(C1=CC=C(C=C1)C(=O)OC(C)C)(O)O
342002-82-8
C10H13BO4
208.02
MFCD02259385
NIQHFKWRHJZMQU-UHFFFAOYSA-N
4-isopropoxycarbonylphenylboronic acid,4-isopropoxycarbonyl phenyl boronic acid,4-isopropoxycarbonyl benzeneboronic acid,propan-2-yl 4-dihydroxyboranyl benzoate,isopropyl 4-boronobenzoate,4-isopropoxycarbonyl phenylboronic acid,p-isopropoxycarbonyl phenylboronic acid,4-isopropoxycarbonylphenylboronicacid,4-propan-2-yloxycarbonylphenyl boronic acid,acmc-209i5o
2734703
(4-propan-2-yloxycarbonylphenyl)boronic acid
B(C1=CC=C(C=C1)C(=O)OC(C)C)(O)O
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Beige
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 208.02
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 4-Isopropoxycarbonylphenylboronic acid, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Recommended Storage : Refrigerator +4°C