Alfa Aesar™ 4-Isopropyl-beta-nitrostyrene, 97%

Manufacturer: Alfa Aesar™ A1095214

Quantity 25g

 View more versions of this product

Catalog No. AAA1095214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 42139-37-7
    Molecular Formula C11H13NO2
    Formula Weight 191.23
    MDL Number MFCD00085100
    Synonym 1-(4-Isopropylphenyl)-2-nitroethylene
    Chemical Name or Material 4-Isopropyl-beta-nitrostyrene

    Specifications

    Formula Weight 191.23
    CAS 42139-37-7
    Molecular Formula C11H13NO2
    MDL Number MFCD00085100
    Melting Point 35° to 38°C
    Quantity 25g
    Synonym 1-(4-Isopropylphenyl)-2-nitroethylene
    Chemical Name or Material 4-Isopropyl-beta-nitrostyrene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit