Alfa Aesar™ 4-(Isopropylamino)benzeneboronic acid, 95%

Manufacturer: Alfa Aesar™ H27800MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27800MD

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 219945-56-9
    Molecular Formula C9H14BNO2
    Formula Weight 179.03
    MDL Number MFCD06798224
    Synonym 4-(Isopropylamino)phenylboronic acid
    Chemical Name or Material 4-(Isopropylamino)benzeneboronic acid

    Specifications

    Formula Weight 179.03
    Molecular Formula C9H14BNO2
    MDL Number MFCD06798224
    CAS 219945-56-9
    Synonym 4-(Isopropylamino)phenylboronic acid
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Melting Point ≈140°C
    Quantity 250mg
    Chemical Name or Material 4-(Isopropylamino)benzeneboronic acid
    Assay 95%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit