Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropylaniline, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
99-88-7
C9H13N
135.21
MFCD00007900
LRTFPLFDLJYEKT-UHFFFAOYSA-N
4-isopropylaniline,cumidine,4-aminocumene,p-isopropylaniline,p-cumidine,4-propan-2-yl aniline,para-isopropylaniline,4-amino-1-isopropylbenzene,cumene, p-amino,aniline, p-isopropyl
7464
CHEBI:43405
4-propan-2-ylaniline
CC(C)C1=CC=C(C=C1)N
99-88-7
C9H13N
135.21
MFCD00007900
LRTFPLFDLJYEKT-UHFFFAOYSA-N
4-isopropylaniline,cumidine,4-aminocumene,p-isopropylaniline,p-cumidine,4-propan-2-yl aniline,para-isopropylaniline,4-amino-1-isopropylbenzene,cumene, p-amino,aniline, p-isopropyl
7464
CHEBI:43405
4-propan-2-ylaniline
CC(C)C1=CC=C(C=C1)N
Specific Gravity 0.989
Flash Point 92°C
Melting Point -63°C
Boiling Point 226°C to 228°C
Refractive Index 1.5420 to 1.544
Infrared Spectrum Authentic
Color Brown to Brown-Red
Packaging Glass bottle
Quantity 10g
Density 0.9890g/mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula (CH3)2CHC6H4NH2
Beilstein 12, 1147
Merck Index 15, 2610
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohols
Formula Weight 135.21
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 4-Isopropylaniline, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-797-2

Recommended Storage : this product may darken on storage