Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropylbenzaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
122-03-2
C10H12O
148.2
MFCD00006953
WTWBUQJHJGUZCY-UHFFFAOYSA-N
4-isopropylbenzaldehyde,cuminaldehyde,cuminic aldehyde,cumaldehyde,cuminal,p-cumic aldehyde,cumic aldehyde,cuminyl aldehyde,p-isopropylbenzaldehyde,benzaldehyde, 4-1-methylethyl
326
CHEBI:28671
4-propan-2-ylbenzaldehyde
CC(C)C1=CC=C(C=C1)C=O
122-03-2
C10H12O
148.2
MFCD00006953
WTWBUQJHJGUZCY-UHFFFAOYSA-N
4-isopropylbenzaldehyde,cuminaldehyde,cuminic aldehyde,cumaldehyde,cuminal,p-cumic aldehyde,cumic aldehyde,cuminyl aldehyde,p-isopropylbenzaldehyde,benzaldehyde, 4-1-methylethyl
326
CHEBI:28671
4-propan-2-ylbenzaldehyde
CC(C)C1=CC=C(C=C1)C=O
Specific Gravity 0.979
Boiling Point 234°C to 235°C
Flash Point 93°C
Refractive Index 1.5290 to 1.532
Quantity 2.5kg
Density 0.9790g/mL
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Merck Index 15, 2611
Solubility Information Solubility in water: insoluble
Formula Weight 148.2
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Isopropylbenzaldehyde, 98%
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 204-516-9

RTECSNumber : CU7000000

TSCA : TSCA