Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropylbenzaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
122-03-2
C10H12O
148.2
MFCD00006953
WTWBUQJHJGUZCY-UHFFFAOYSA-N
4-isopropylbenzaldehyde,cuminaldehyde,cuminic aldehyde,cumaldehyde,cuminal,p-cumic aldehyde,cumic aldehyde,cuminyl aldehyde,p-isopropylbenzaldehyde,benzaldehyde, 4-1-methylethyl
326
CHEBI:28671
4-propan-2-ylbenzaldehyde
CC(C)C1=CC=C(C=C1)C=O
122-03-2
C10H12O
148.2
MFCD00006953
WTWBUQJHJGUZCY-UHFFFAOYSA-N
4-isopropylbenzaldehyde,cuminaldehyde,cuminic aldehyde,cumaldehyde,cuminal,p-cumic aldehyde,cumic aldehyde,cuminyl aldehyde,p-isopropylbenzaldehyde,benzaldehyde, 4-1-methylethyl
326
CHEBI:28671
4-propan-2-ylbenzaldehyde
CC(C)C1=CC=C(C=C1)C=O
Boiling Point 234°C to 235°C
Flash Point 93°C
Specific Gravity 0.979
Refractive Index 1.5290 to 1.532
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless to Yellow
Density 0.9790g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Merck Index 15, 2611
Solubility Information Solubility in water: insoluble
Formula Weight 148.2
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 4-Isopropylbenzaldehyde, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 204-516-9

RTECSNumber : CU7000000

TSCA : TSCA