Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 4-Isopropylbenzenesulfonyl chloride, 96%
Click to view available options
Quantity:
5g
25g
Specifications
Sensitivity | Moisture sensitive |
Refractive Index | 1.539 |
Density | 1.22 |
Flash Point | >110°C (230°F) |
Boiling Point | 142°C (5mmHg) |
Quantity | 5g |
UN Number | UN3265 |
Beilstein | 1103862 |
Formula Weight | 218.7 |
Percent Purity | 96% |
Show More |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures