Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Isopropylthiazole-2-carboxylic acid, 97%

Click to view available options
Quantity:
5g
300831-06-5
C7H9NO2S
171.214
MFCD09864275
RDWHKWXYJQUZNS-UHFFFAOYSA-N
4-isopropylthiazole-2-carboxylic acid,2-thiazolecarboxylic acid, 4-1-methylethyl,4-isopropyl-1,3-thiazole-2-carboxylic acid,4-1-methylethyl-2-thiazolecarboxylicacid,4-isopropyl-thiazole-2-carboxylic acid,4-propan-2-yl-1,3-thiazole-2-carboxylic acid,4-1-methylethyl-2-thiazolecarboxylic acid,pubchem19253,4-isopropylthiazole-2-carboxlic acid,4-isopropyl-thiazole-2-carboxlic acid
22499307
4-propan-2-yl-1,3-thiazole-2-carboxylic acid
CC(C)C1=CSC(=N1)C(=O)O
300831-06-5
C7H9NO2S
171.214
MFCD09864275
RDWHKWXYJQUZNS-UHFFFAOYSA-N
4-isopropylthiazole-2-carboxylic acid,2-thiazolecarboxylic acid, 4-1-methylethyl,4-isopropyl-1,3-thiazole-2-carboxylic acid,4-1-methylethyl-2-thiazolecarboxylicacid,4-isopropyl-thiazole-2-carboxylic acid,4-propan-2-yl-1,3-thiazole-2-carboxylic acid,4-1-methylethyl-2-thiazolecarboxylic acid,pubchem19253,4-isopropylthiazole-2-carboxlic acid,4-isopropyl-thiazole-2-carboxlic acid
22499307
4-propan-2-yl-1,3-thiazole-2-carboxylic acid
CC(C)C1=CSC(=N1)C(=O)O
Quantity 5g
Beilstein 14226164
Formula Weight 171.22
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 4-Isopropylthiazole-2-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning