Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Mercapto-1H-pyrazolo[3,4-d]pyrimidine, 98%, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
5334-23-6
C5H4N4S
152.175
MFCD00005687
PYAOPMWCFSVFOT-UHFFFAOYSA-N
tisopurine,thiopurinol,1h-pyrazolo 3,4-d pyrimidine-4-thiol,allopurinol, thio,mpp pharmaceutical,4-mercaptopyrazolo 3,4-d pyrimidine,4-mercapto-1h-pyrazolo 3,4-d pyrimidine,tisopurine inn,unii-79f9i2r16m
667510
1,2-dihydropyrazolo[3,4-d]pyrimidine-4-thione
C1=C2C(=NC=NC2=S)NN1
5334-23-6
C5H4N4S
152.175
MFCD00005687
PYAOPMWCFSVFOT-UHFFFAOYSA-N
tisopurine,thiopurinol,1h-pyrazolo 3,4-d pyrimidine-4-thiol,allopurinol, thio,mpp pharmaceutical,4-mercaptopyrazolo 3,4-d pyrimidine,4-mercapto-1h-pyrazolo 3,4-d pyrimidine,tisopurine inn,unii-79f9i2r16m
667510
1,2-dihydropyrazolo[3,4-d]pyrimidine-4-thione
C1=C2C(=NC=NC2=S)NN1
Quantity 5g
Packaging Glass bottle
Melting Point ~355°C (decomposition)
Formula Weight 152.18
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 4-Mercapto-1H-pyrazolo[3,4-d]pyrimidine