Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Mercaptophenol, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
637-89-8
C6H6OS
126.18
MFCD00004850
BXAVKNRWVKUTLY-UHFFFAOYSA-N
4-mercaptophenol,4-hydroxythiophenol,4-hydroxybenzenethiol,p-hydroxythiophenol,phenol, 4-mercapto,p-mercaptophenol,thiohydroquinone,monothiohydroquinone,4-hydroxy thiophenol,hydroquinone, monothio
240147
4-sulfanylphenol
C1=CC(=CC=C1O)S
637-89-8
C6H6OS
126.18
MFCD00004850
BXAVKNRWVKUTLY-UHFFFAOYSA-N
4-mercaptophenol,4-hydroxythiophenol,4-hydroxybenzenethiol,p-hydroxythiophenol,phenol, 4-mercapto,p-mercaptophenol,thiohydroquinone,monothiohydroquinone,4-hydroxy thiophenol,hydroquinone, monothio
240147
4-sulfanylphenol
C1=CC(=CC=C1O)S
Melting Point 33°C to 35°C
Boiling Point 149°C to 150°C (25.0mmHg)
Flash Point 110°C
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula SHC6H4OH
Beilstein 06, IV, 5790
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,alkali solution,concentrated,sulfuric acid
Formula Weight 126.18
Physical Form Liquid or Low Melting Solid
Percent Purity 99%
Chemical Name or Material 4-Mercaptophenol, 99%
Show More Show Less

GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously

GHS Signal Word: Danger

EINECSNumber : 211-307-6