Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methanesulfonamido)phenylboronic acid, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
380430-57-9
C7H10BNO4S
215.04
NDVJJEADFLTFCD-UHFFFAOYSA-N
4-methylsulfonylamino phenylboronic acid,4-methylsulfonamido phenylboronic acid,4-methanesulfonylaminophenylboronic acid,4-methylsulfonylaminophenylboronic acid,4-methanesulfonylamino phenylboronic acid,4-methylsulfonamido phenyl boronic acid,4-methanesulfonamidophenyl boronic acid,4-methanesulfonamido phenyl boronic acid,4-methylsulphonyl amino benzeneboronic acid,4-methylsulfonylamino benzeneboronic acid
2773537
[4-(methanesulfonamido)phenyl]boronic acid
B(C1=CC=C(C=C1)NS(=O)(=O)C)(O)O
380430-57-9
C7H10BNO4S
215.04
NDVJJEADFLTFCD-UHFFFAOYSA-N
4-methylsulfonylamino phenylboronic acid,4-methylsulfonamido phenylboronic acid,4-methanesulfonylaminophenylboronic acid,4-methylsulfonylaminophenylboronic acid,4-methanesulfonylamino phenylboronic acid,4-methylsulfonamido phenyl boronic acid,4-methanesulfonamidophenyl boronic acid,4-methanesulfonamido phenyl boronic acid,4-methylsulphonyl amino benzeneboronic acid,4-methylsulfonylamino benzeneboronic acid
2773537
[4-(methanesulfonamido)phenyl]boronic acid
B(C1=CC=C(C=C1)NS(=O)(=O)C)(O)O
Melting Point 234°C to 236°C
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 96% min. (HPLC)
Formula Weight 215.04
Percent Purity 97%
Chemical Name or Material 4-(Methanesulfonamido)phenylboronic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning