Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-2-methylaniline 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
102-50-1
C8H11NO
137.182
MFCD00007735
CDGNLUSBENXDGG-UHFFFAOYSA-N
m-cresidine,2-methyl-4-methoxyaniline,2-methyl-p-anisidine,4-methoxy-o-toluidine,benzenamine, 4-methoxy-2-methyl,meta-cresidine,p-anisidine, 2-methyl,6-amino-3-methoxytoluene,2-methyl-4-methoxybenzenamine,4-methoxy-2-methylbenzenamine
7610
CHEBI:82430
4-methoxy-2-methylaniline
CC1=C(C=CC(=C1)OC)N
102-50-1
C8H11NO
137.182
MFCD00007735
CDGNLUSBENXDGG-UHFFFAOYSA-N
m-cresidine,2-methyl-4-methoxyaniline,2-methyl-p-anisidine,4-methoxy-o-toluidine,benzenamine, 4-methoxy-2-methyl,meta-cresidine,p-anisidine, 2-methyl,6-amino-3-methoxytoluene,2-methyl-4-methoxybenzenamine,4-methoxy-2-methylbenzenamine
7610
CHEBI:82430
4-methoxy-2-methylaniline
CC1=C(C=CC(=C1)OC)N
Infrared Spectrum Authentic
Density 1.0600g/mL
Refractive Index 1.5670 to 1.5700 (20°C, 589nm)
Quantity 5g
Packaging Glass bottle
Color Black to Brown
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3OC6H3(CH3)NH2
Beilstein 13, II, 330
Formula Weight 137.18
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 4-Methoxy-2-methylaniline, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Suspected of causing cancer.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-036-7

RTECSNumber : BZ6730000

TSCA : TSCA