Alfa Aesar™ 4-Methoxy-2-nitroaniline, 99%

Manufacturer: Alfa Aesar™ A1167336

Quantity 500g

 View more versions of this product

Catalog No. AAA1167336

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 96-96-8
    Molecular Formula C7H8N2O3
    Formula Weight 168.15
    MDL Number MFCD00007152
    UN Number UN2811
    Beilstein 880318
    Synonym 4-Amino-3-nitroanisole; 2-Nitro-p-anisidine
    Chemical Name or Material 4-Methoxy-2-nitroaniline

    Specifications

    Formula Weight 168.15
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 96-96-8
    Synonym 4-Amino-3-nitroanisole; 2-Nitro-p-anisidine
    Quantity 500g
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    TSCA TSCA
    UN Number UN2811
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H373
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    RTECS Number BY4415000
    Molecular Formula C7H8N2O3
    MDL Number MFCD00007152
    Beilstein 880318
    Melting Point 126° to 128°C
    Chemical Name or Material 4-Methoxy-2-nitroaniline
    Assay 99%

    Description

    Applications
    4-Methoxy-2-nitroaniline is an aniline compound used in the synthesis of dyes and pigments. It has also been shown to act as a molluscicidal agent and has been used in hemolytic anemia studies. Also used as an intermediate for the synthesis of dyes, pigments and other chemical compounds.

    Solubility
    Slightly soluble in water. Soluble in hot methanol almost transparency.

    Notes
    Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.

    GHS H Statement
    H300-H310-H330-H373
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit