Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methoxy-2-(trifluoromethoxy)benzoic acid, 97%

Click to view available options
Quantity:
1g
5g
886502-37-0
C9H7F3O4
236.146
MFCD06660285
PDDVYCWKVQXOTR-UHFFFAOYSA-N
4-methoxy-2-trifluoromethoxy benzoic acid
17750721
4-methoxy-2-(trifluoromethoxy)benzoic acid
COC1=CC(=C(C=C1)C(=O)O)OC(F)(F)F
886502-37-0
C9H7F3O4
236.146
MFCD06660285
PDDVYCWKVQXOTR-UHFFFAOYSA-N
4-methoxy-2-trifluoromethoxy benzoic acid
17750721
4-methoxy-2-(trifluoromethoxy)benzoic acid
COC1=CC(=C(C=C1)C(=O)O)OC(F)(F)F
Melting Point 99°C to 101°C
Quantity 1g
Formula Weight 236.15
Percent Purity 97%
Chemical Name or Material 4-Methoxy-2-(trifluoromethoxy)benzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures