Alfa Aesar™ 4-Methoxy-2-(trifluoromethyl)benzenesulfonyl chloride, 97%

Manufacturer: Alfa Aesar™ H3228003

Quantity 1g

 View more versions of this product

Catalog No. AAH3228003

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C8H6ClF3O3S
    Formula Weight 274.64
    MDL Number MFCD11519323
    UN Number UN3261
    Chemical Name or Material 4-Methoxy-2-(trifluoromethyl)benzenesulfonyl chloride

    Specifications

    Formula Weight 274.64
    MDL Number MFCD11519323
    UN Number UN3261
    Molecular Formula C8H6ClF3O3S
    Melting Point 57° to 60°C
    Quantity 1g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 4-Methoxy-2-(trifluoromethyl)benzenesulfonyl chloride
    Assay 97%

    Description

    Applications
    4-Methoxy-2-(trifluoromethyl)benzenesulfonyl chloride, is used as a pharmaceutical intermediate.

    Solubility
    Insoluble in water.

    Notes
    Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit