Alfa Aesar™ 4-Methoxy-3,5-dimethylaniline, 98%

Manufacturer: Alfa Aesar™ H32232MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH32232MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 39785-37-0
    Molecular Formula C9H13NO
    Formula Weight 151.21
    MDL Number MFCD01907075
    UN Number UN2811
    Synonym 4-Amino-2,6-dimethylanisole
    Chemical Name or Material 4-Methoxy-3,5-dimethylaniline

    Specifications

    Formula Weight 151.21
    Molecular Formula C9H13NO
    MDL Number MFCD01907075
    CAS 39785-37-0
    UN Number UN2811
    Synonym 4-Amino-2,6-dimethylanisole
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H311-H302-H332-H315-H319
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Melting Point 62° to 65°C
    Quantity 250mg
    Chemical Name or Material 4-Methoxy-3,5-dimethylaniline
    Assay 98%

    GHS H Statement
    H311-H302-H332-H315-H319
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit