Learn More
4-Methoxy-3-buten-2-one, tech. 90%, Thermo Scientific Chemicals
Description
4-Methoxy-3-buten-2-one is an important precursor for the synthesis of 1-methoxy-3-trimethylsilyloxy-1,3-butadiene and the Danishefsky diene and its equivalent. It is widely used as a reactant in the synthesis of hydroisoquinoline derivatives. It is employed as an intermediate for the synthesis of manzamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Methoxy-3-buten-2-one is an important precursor for the synthesis of 1-methoxy-3-trimethylsilyloxy-1,3-butadiene and the Danishefsky diene and its equivalent. It is widely used as a reactant in the synthesis of hydroisoquinoline derivatives. It is employed as an intermediate for the synthesis of manzamine.
Solubility
Miscible with tetrahydrofuran, ether, acetonitrile and benzene.
Notes
Keep away from sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
Specifications
Density | 0.99 |
Boiling Point | 66°C to 69°C (12 mmHg) |
Flash Point | 63°C (145°F) |
Refractive Index | 1.465 |
Quantity | 250 g |
Beilstein | 1741053 |
Solubility Information | Miscible with tetrahydrofuran,ether,acetonitrile and benzene. |
Formula Weight | 100.12 |
Percent Purity | 90% |
Grade | Technical |
Show More |
Safety and Handling
Hazard Category | H227-H315-H319-H335 |
Hazard Statement | GHS H Statement H227 Combustible liquid. |
Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
EINECSNumber | 225-087-4 |
TSCA | No |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.