Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-3-methylphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
175883-62-2
C8H11BO3
165.99
MFCD02179464
PXVDQGVAZBTFIB-UHFFFAOYSA-N
4-methoxy-3-methylbenzeneboronic acid,4-methoxy-3-methylphenyl boronic acid,4-methoxy-3-methylphenyl boranediol,3-methyl-4-methoxyphenylboronic acid,boronic acid, 4-methoxy-3-methylphenyl,boronic acid,b-4-methoxy-3-methylphenyl,b-4-methoxy-3-methylphenyl boronic acid,akos brn-0220,4-methoxy-3-methyl-phenyl boronic acid,timtec-bb sbb005903
2773487
(4-methoxy-3-methylphenyl)boronic acid
B(C1=CC(=C(C=C1)OC)C)(O)O
175883-62-2
C8H11BO3
165.99
MFCD02179464
PXVDQGVAZBTFIB-UHFFFAOYSA-N
4-methoxy-3-methylbenzeneboronic acid,4-methoxy-3-methylphenyl boronic acid,4-methoxy-3-methylphenyl boranediol,3-methyl-4-methoxyphenylboronic acid,boronic acid, 4-methoxy-3-methylphenyl,boronic acid,b-4-methoxy-3-methylphenyl,b-4-methoxy-3-methylphenyl boronic acid,akos brn-0220,4-methoxy-3-methyl-phenyl boronic acid,timtec-bb sbb005903
2773487
(4-methoxy-3-methylphenyl)boronic acid
B(C1=CC(=C(C=C1)OC)C)(O)O
Color Purple to White
Quantity 10g
Melting Point 219.3°C to 220.8°C
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 165.99
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-methylphenylboronic acid
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning