Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-3-nitrobenzoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
89-41-8
C8H7NO5
197.15
MFCD00007256
ANXBDAFDZSXOPQ-UHFFFAOYSA-N
4-methoxy-3-nitrobenzoic acid, 3-nitro-4-methoxybenzoic acid, 3-nitro-p-anisic acid, benzoic acid, 4-methoxy-3-nitro, unii-dd71o31olz, p-anisic acid, 3-nitro, dd71o31olz, 4-methoxy-3-nitro benzoic acid, 4-methoxy-3-nitro-benzoic acid, 4-methoxy-3-nitrobenzoicacid
66640
COC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]
89-41-8
C8H7NO5
197.15
MFCD00007256
ANXBDAFDZSXOPQ-UHFFFAOYSA-N
4-methoxy-3-nitrobenzoic acid, 3-nitro-4-methoxybenzoic acid, 3-nitro-p-anisic acid, benzoic acid, 4-methoxy-3-nitro, unii-dd71o31olz, p-anisic acid, 3-nitro, dd71o31olz, 4-methoxy-3-nitro benzoic acid, 4-methoxy-3-nitro-benzoic acid, 4-methoxy-3-nitrobenzoicacid
66640
COC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 189°C to 194°C
Color Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3OC6H3(NO2)CO2H
Beilstein 10, 181
Formula Weight 197.15
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Methoxy-3-nitrobenzoic acid, 98%
Show More Show Less

EINECSNumber : 201-906-0

TSCA : TSCA