Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methoxy-3-nitropyridine, 98%

Click to view available options
Quantity:
1g
5g
31872-62-5
C6H6N2O3
154.125
MFCD00209661
BZPVREXVOZITPF-UHFFFAOYSA-N
4-methoxy-3-nitro-pyridine,pyridine, 4-methoxy-3-nitro,methyl 3-nitro-4-pyridinyl ether,3-nitro-4-methoxypyridine,methyl3-nitro-4-pyridinylether,pubchem14990,acmc-1ctwk,3-nitro-4-methoxy pyridine,ksc494q6d,#
355832
4-methoxy-3-nitropyridine
COC1=C(C=NC=C1)[N+](=O)[O-]
31872-62-5
C6H6N2O3
154.125
MFCD00209661
BZPVREXVOZITPF-UHFFFAOYSA-N
4-methoxy-3-nitro-pyridine,pyridine, 4-methoxy-3-nitro,methyl 3-nitro-4-pyridinyl ether,3-nitro-4-methoxypyridine,methyl3-nitro-4-pyridinylether,pubchem14990,acmc-1ctwk,3-nitro-4-methoxy pyridine,ksc494q6d,#
355832
4-methoxy-3-nitropyridine
COC1=C(C=NC=C1)[N+](=O)[O-]
Quantity 1g
Formula Weight 154.13
Percent Purity 98%
Chemical Name or Material 4-Methoxy-3-nitropyridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning