Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-3-pyridineboronic acid hydrate, 97%, Acros Organics

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
1266615-79-5,355004-67-0
C6H8BNO3·xH2O
152.94
MFCD07186213
YUTPAZKVEOJQCY-UHFFFAOYSA-N
4-methoxy-3-pyridineboronic acid,4-methoxypyridin-3-yl boronic acid,4-methoxypyridine-3-boronic acid,3-borono-4-methoxypyridine,4-methoxy-3-pyridylboronic acid,boronic acid, 4-methoxy-3-pyridinyl,4-methoxy-pyridine-3-boric acid,acmc-209iha,4-methoxy-5-pyridineboronic acid,4-methoxy-pyridin-3-boronic acid
2762556
(4-methoxypyridin-3-yl)boronic acid
B(C1=C(C=CN=C1)OC)(O)O
1266615-79-5,355004-67-0
C6H8BNO3·xH2O
152.94
MFCD07186213
YUTPAZKVEOJQCY-UHFFFAOYSA-N
4-methoxy-3-pyridineboronic acid,4-methoxypyridin-3-yl boronic acid,4-methoxypyridine-3-boronic acid,3-borono-4-methoxypyridine,4-methoxy-3-pyridylboronic acid,boronic acid, 4-methoxy-3-pyridinyl,4-methoxy-pyridine-3-boric acid,acmc-209iha,4-methoxy-5-pyridineboronic acid,4-methoxy-pyridin-3-boronic acid
2762556
(4-methoxypyridin-3-yl)boronic acid
B(C1=C(C=CN=C1)OC)(O)O
Packaging Glass bottle
Infrared Spectrum Authentic
Color Cream
Quantity 250mg
Assay Percent Range 96% min. (HPLC)
Formula Weight 152.94
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-pyridineboronic acid hydrate

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning