Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-3-(trifluoromethyl)aniline, 98+%, Alfa Aesar™

Click to view available options
Quantity:
2g
10g
393-15-7
C8H8F3NO
191.153
MFCD00043460
CQJCPOVTPNWVBW-UHFFFAOYSA-N
4-methoxy-3-trifluoromethyl aniline,5-amino-2-methoxybenzotrifluoride,3-trifluoromethyl-p-anisidine,3-amino-6-methoxybenzotrifluoride,4-methoxy-3-trifluoromethyl-phenylamine,benzenamine, 4-methoxy-3-trifluoromethyl,4-methoxy-3-trifluoromethyl phenylamine,acmc-209j4q,3-amino-6-methoxybenzotrifloride
600705
4-methoxy-3-(trifluoromethyl)aniline
COC1=C(C=C(C=C1)N)C(F)(F)F
393-15-7
C8H8F3NO
191.153
MFCD00043460
CQJCPOVTPNWVBW-UHFFFAOYSA-N
4-methoxy-3-trifluoromethyl aniline,5-amino-2-methoxybenzotrifluoride,3-trifluoromethyl-p-anisidine,3-amino-6-methoxybenzotrifluoride,4-methoxy-3-trifluoromethyl-phenylamine,benzenamine, 4-methoxy-3-trifluoromethyl,4-methoxy-3-trifluoromethyl phenylamine,acmc-209j4q,3-amino-6-methoxybenzotrifloride
600705
4-methoxy-3-(trifluoromethyl)aniline
COC1=C(C=C(C=C1)N)C(F)(F)F
Melting Point 57°C to 61°C
Quantity 2g
Boiling Point 112°C (18mmHg)
UN Number UN2811
Beilstein 2723972
Formula Weight 191.15
Percent Purity ≥98%
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)aniline

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.