Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methoxy-3-(trifluoromethyl)benzoic acid, 97%

Click to view available options
Quantity:
1g
5g
213598-09-5
C9H7F3O3
220.147
MFCD01091014
VNWUPESTVBTYJM-UHFFFAOYSA-N
4-methoxy-3-trifluoromethyl benzoic acid,4-methoxy-3-trifluoromethyl-benzoic acid,acmc-1cr7h,3-trifluoromethyl-4-methoxybenzoic acid,benzoic acid, 4-methoxy-3-trifluoromethyl,4-methyloxy-3-trifluoromethyl benzoic acid
2775295
4-methoxy-3-(trifluoromethyl)benzoic acid
COC1=C(C=C(C=C1)C(=O)O)C(F)(F)F
213598-09-5
C9H7F3O3
220.147
MFCD01091014
VNWUPESTVBTYJM-UHFFFAOYSA-N
4-methoxy-3-trifluoromethyl benzoic acid,4-methoxy-3-trifluoromethyl-benzoic acid,acmc-1cr7h,3-trifluoromethyl-4-methoxybenzoic acid,benzoic acid, 4-methoxy-3-trifluoromethyl,4-methyloxy-3-trifluoromethyl benzoic acid
2775295
4-methoxy-3-(trifluoromethyl)benzoic acid
COC1=C(C=C(C=C1)C(=O)O)C(F)(F)F
Melting Point 169°C to 171°C
Quantity 1g
Beilstein 8619237
Formula Weight 220.15
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning