Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methoxy-3-(trifluoromethyl)benzonitrile, 97+%

Click to view available options
Quantity:
1g
5g
261951-87-5
C9H6F3NO
201.148
MFCD01631575
FLHQJZPLFRIGOT-UHFFFAOYSA-N
4-methoxy-3-trifluoromethyl benzonitrile,5-cyano-2-methoxybenzotrifluoride,4-methoxy-3-trifluoromethyl-benzonitrile,4-methoxy-3-trifluoromethyl benzenecarbonitrile,acmc-20a3lw,4-cyano-2-trifluoromethyl anisole,3-trifluoromethyl-p-anisonitrile,5-cyano-2-trifluoromethyl anisole,benzonitrile,4-methoxy-3-trifluoromethyl
2775296
4-methoxy-3-(trifluoromethyl)benzonitrile
COC1=C(C=C(C=C1)C#N)C(F)(F)F
261951-87-5
C9H6F3NO
201.148
MFCD01631575
FLHQJZPLFRIGOT-UHFFFAOYSA-N
4-methoxy-3-trifluoromethyl benzonitrile,5-cyano-2-methoxybenzotrifluoride,4-methoxy-3-trifluoromethyl-benzonitrile,4-methoxy-3-trifluoromethyl benzenecarbonitrile,acmc-20a3lw,4-cyano-2-trifluoromethyl anisole,3-trifluoromethyl-p-anisonitrile,5-cyano-2-trifluoromethyl anisole,benzonitrile,4-methoxy-3-trifluoromethyl
2775296
4-methoxy-3-(trifluoromethyl)benzonitrile
COC1=C(C=C(C=C1)C#N)C(F)(F)F
Melting Point 98°C to 100°C
Quantity 1g
UN Number UN3439
Formula Weight 201.15
Percent Purity ≥97%
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzonitrile

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.