Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-4'-fluorochalcone, 98%, Acros Organics

Click to view available options
Quantity:
5g
2965-64-2
C16H13FO2
256.276
MFCD00017964
AOINTYAZKYYNGN-NYYWCZLTSA-N
4-methoxy-4'-fluorochalcone,4'-fluoro-4-methoxychalcone,1-4-fluorophenyl-3-4-methoxyphenyl prop-2-en-1-one,e-1-4-fluorophenyl-3-4-methoxyphenyl prop-2-en-1-one,e-1-4-fluorophenyl-3-4-methoxyphenyl-2-propen-1-one,2e-1-4-fluorophenyl-3-4-methoxyphenyl prop-2-en-1-one,e-4'-fluoro-4-methoxychalcone,trans-4'-fluoro-4-methoxychalcone,2e-1-4-fluorophenyl-3-4-methoxyphenyl-2-propen-1-one #
5377456
(E)-1-(4-fluorophenyl)-3-(4-methoxyphenyl)prop-2-en-1-one
COC1=CC=C(C=C1)C=CC(=O)C2=CC=C(C=C2)F
2965-64-2
C16H13FO2
256.276
MFCD00017964
AOINTYAZKYYNGN-NYYWCZLTSA-N
4-methoxy-4'-fluorochalcone,4'-fluoro-4-methoxychalcone,1-4-fluorophenyl-3-4-methoxyphenyl prop-2-en-1-one,e-1-4-fluorophenyl-3-4-methoxyphenyl prop-2-en-1-one,e-1-4-fluorophenyl-3-4-methoxyphenyl-2-propen-1-one,2e-1-4-fluorophenyl-3-4-methoxyphenyl prop-2-en-1-one,e-4'-fluoro-4-methoxychalcone,trans-4'-fluoro-4-methoxychalcone,2e-1-4-fluorophenyl-3-4-methoxyphenyl-2-propen-1-one #
5377456
(E)-1-(4-fluorophenyl)-3-(4-methoxyphenyl)prop-2-en-1-one
COC1=CC=C(C=C1)C=CC(=O)C2=CC=C(C=C2)F
Quantity 5g
Melting Point 103°C to 108°C
Infrared Spectrum Authentic
Color Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 256.28
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 4-Methoxy-4'-fluorochalcone
Show More Show Less