Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-7-methyl-5H-furo[3,2-g][1]benzopyran-5-one, 97%, Thermo Scientific Chemicals

Catalog Number ac235030050
Click to view available options
Quantity:
5 g
25 g
82-57-5
MFCD00005008
NZVQLVGOZRELTG-UHFFFAOYSA-N
visnagin, desmethoxykhellin, visnagidin, visnagine, 5-methoxy-2-methylfuranochromone, 4-methoxy-7-methyl-5h-furo 3,2-g chromen-5-one, unii-p64438mlbw, 4-methoxy-7-methyl-furo 3,2-g chromen-5-one, chembl45176, 4-methoxy-7-methyl-5h-furo 3,2-g 1-benzopyran-5-one
6716
CHEBI:10002
4-methoxy-7-methylfuro[3,2-g]chromen-5-one
CC1=CC(=O)C2=C(O1)C=C3C(=C2OC)C=CO3
82-57-5
MFCD00005008
NZVQLVGOZRELTG-UHFFFAOYSA-N
visnagin, desmethoxykhellin, visnagidin, visnagine, 5-methoxy-2-methylfuranochromone, 4-methoxy-7-methyl-5h-furo 3,2-g chromen-5-one, unii-p64438mlbw, 4-methoxy-7-methyl-furo 3,2-g chromen-5-one, chembl45176, 4-methoxy-7-methyl-5h-furo 3,2-g 1-benzopyran-5-one
6716
CHEBI:10002
4-methoxy-7-methylfuro[3,2-g]chromen-5-one
CC1=CC(=O)C2=C(O1)C=C3C(=C2OC)C=CO3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 139°C to 142°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C13H10O4
Quantity 5 g
Merck Index 15, 10209
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in chloroform,sparingly soluble in alcohol
Molecular Weight (g/mol) 230.22
Formula Weight 230.22
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Methoxy-7-methyl-5H-furo[3, 2-g][1]benzopyran-5-one, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-430-3
RTECSNumber LV1420000

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit