Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-alpha-toluenethiol, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
6258-60-2
C8H10OS
154.23
MFCD00004871
PTDVPWWJRCOIIO-UHFFFAOYSA-N
4-methoxybenzyl mercaptan,4-methoxy-alpha-toluenethiol,4-methoxybenzylmercaptan,4-methoxyphenyl methanethiol,4-methoxy benzylmercaptan,4-methoxybenzenemethanethiol,benzenemethanethiol, 4-methoxy,p-methoxybenzyl mercaptan,p-methoxybenzylthiol,p-methoxybenzylmercaptan
80407
(4-methoxyphenyl)methanethiol
COC1=CC=C(C=C1)CS
6258-60-2
C8H10OS
154.23
MFCD00004871
PTDVPWWJRCOIIO-UHFFFAOYSA-N
4-methoxybenzyl mercaptan,4-methoxy-alpha-toluenethiol,4-methoxybenzylmercaptan,4-methoxyphenyl methanethiol,4-methoxy benzylmercaptan,4-methoxybenzenemethanethiol,benzenemethanethiol, 4-methoxy,p-methoxybenzyl mercaptan,p-methoxybenzylthiol,p-methoxybenzylmercaptan
80407
(4-methoxyphenyl)methanethiol
COC1=CC=C(C=C1)CS
Specific Gravity 1.107
Flash Point >110°C
Boiling Point 90°C to 95°C (0.5mmHg)
Refractive Index 1.5720 to 1.578
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless to Yellow
Density 1.1070g/mL
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula CH3OC6H4CH2SH
Beilstein 06, 901
Solubility Information Solubility in water: insoluble
Formula Weight 154.23
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 4-Methoxy-α-toluenethiol
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Danger

EINECSNumber : 228-393-6