Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-alpha-toluenethiol, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
6258-60-2
C8H10OS
154.23
MFCD00004871
PTDVPWWJRCOIIO-UHFFFAOYSA-N
4-methoxybenzyl mercaptan,4-methoxy-alpha-toluenethiol,4-methoxybenzylmercaptan,4-methoxyphenyl methanethiol,4-methoxy benzylmercaptan,4-methoxybenzenemethanethiol,benzenemethanethiol, 4-methoxy,p-methoxybenzyl mercaptan,p-methoxybenzylthiol,p-methoxybenzylmercaptan
80407
(4-methoxyphenyl)methanethiol
COC1=CC=C(C=C1)CS
6258-60-2
C8H10OS
154.23
MFCD00004871
PTDVPWWJRCOIIO-UHFFFAOYSA-N
4-methoxybenzyl mercaptan,4-methoxy-alpha-toluenethiol,4-methoxybenzylmercaptan,4-methoxyphenyl methanethiol,4-methoxy benzylmercaptan,4-methoxybenzenemethanethiol,benzenemethanethiol, 4-methoxy,p-methoxybenzyl mercaptan,p-methoxybenzylthiol,p-methoxybenzylmercaptan
80407
(4-methoxyphenyl)methanethiol
COC1=CC=C(C=C1)CS
Boiling Point 90°C to 95°C (0.5mmHg)
Flash Point >110°C
Specific Gravity 1.107
Refractive Index 1.5720 to 1.578
Infrared Spectrum Authentic
Density 1.1070g/mL
Color Colorless to Yellow
Packaging Glass bottle
Quantity 100g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula CH3OC6H4CH2SH
Beilstein 06, 901
Solubility Information Solubility in water: insoluble
Formula Weight 154.23
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material 4-Methoxy-α-toluenethiol
Show More Show Less

GHS H Statement:
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger