Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methoxy-D-phenylalanine, 95%

Click to view available options
Quantity:
1g
5g
25g
39878-65-4
C10H13NO3
195.218
MFCD00190961
GEYBMYRBIABFTA-SECBINFHSA-N
o-methyl-d-tyrosine,4-methoxy-d-phenylalanine,h-d-tyr me-oh,r-2-amino-3-4-methoxyphenyl propanoic acid,d-tyrosine, o-methyl,2r-2-amino-3-4-methoxyphenyl propanoic acid,r-2-amino-3-4-methoxyphenyl propionic acid,h-4-methoxy-d-phe-oh,o-methyl-d-tyrosine nt
6950205
(2R)-2-amino-3-(4-methoxyphenyl)propanoic acid
COC1=CC=C(C=C1)CC(C(=O)O)N
39878-65-4
C10H13NO3
195.218
MFCD00190961
GEYBMYRBIABFTA-SECBINFHSA-N
o-methyl-d-tyrosine,4-methoxy-d-phenylalanine,h-d-tyr me-oh,r-2-amino-3-4-methoxyphenyl propanoic acid,d-tyrosine, o-methyl,2r-2-amino-3-4-methoxyphenyl propanoic acid,r-2-amino-3-4-methoxyphenyl propionic acid,h-4-methoxy-d-phe-oh,o-methyl-d-tyrosine nt
6950205
(2R)-2-amino-3-(4-methoxyphenyl)propanoic acid
COC1=CC=C(C=C1)CC(C(=O)O)N
Beilstein 3201704
Formula Weight 195.22
Physical Form Solid
Percent Purity 95%
Quantity 25g
Chemical Name or Material 4-Methoxy-D-phenylalanine

storageNote1 : 2° to 8°C