Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methoxy-N-methylaniline, 96%

Click to view available options
Quantity:
1g
5g
25g
5961-59-1
C8H11NO
137.182
MFCD00008399
JFXDIXYFXDOZIT-UHFFFAOYSA-N
n-methyl-p-anisidine,n-methyl-4-anisidine,n-methyl-4-methoxyaniline,benzenamine, 4-methoxy-n-methyl,n-methyl-p-ansidine,p-anisidine, n-methyl,4-methoxy-n-methylbenzenamine,1-methylamino-4-methoxybenzene,p-methoxy-n-methylaniline,acmc-209mes
22250
4-methoxy-N-methylaniline
CNC1=CC=C(C=C1)OC
5961-59-1
C8H11NO
137.182
MFCD00008399
JFXDIXYFXDOZIT-UHFFFAOYSA-N
n-methyl-p-anisidine,n-methyl-4-anisidine,n-methyl-4-methoxyaniline,benzenamine, 4-methoxy-n-methyl,n-methyl-p-ansidine,p-anisidine, n-methyl,4-methoxy-n-methylbenzenamine,1-methylamino-4-methoxybenzene,p-methoxy-n-methylaniline,acmc-209mes
22250
4-methoxy-N-methylaniline
CNC1=CC=C(C=C1)OC
Odor Odorless
Color Yellow
Melting Point 33°C to 36°C
Boiling Point 135°C to 136°C (19mmHg)
Quantity 25g
UN Number UN2811
Beilstein 774640
Formula Weight 137.18
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 4-Methoxy-N-methylaniline
Show More Show Less

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.