Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-o-phenylenediamine dihydrochloride, 97+%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
25g
59548-39-9
C7H12Cl2N2O
211.086
MFCD00052003
SXCHMHOBHJOXGC-UHFFFAOYSA-N
4-methoxybenzene-1,2-diamine dihydrochloride, 4-methoxy-o-phenylenediamine dihydrochloride, 3,4-diaminoanisole dihydrochloride, 1,2-benzenediamine, 4-methoxy-, dihydrochloride, 3,4-diaminoanisole 2hcl, 4-methoxy-phenylenediamine dihydrochloride, 4-methoxy-1,2-benzenediamine dihydrochloride, 4-methoxy-benzene-1,2-diamine dihydrochloride, 4-methoxy-1,2-phenylenediamine dihydrochloride, 4-methoxy-l,2-phenylene diamine dihydrochloride
3085161
4-methoxybenzene-1,2-diamine;dihydrochloride
COC1=CC(=C(C=C1)N)N.Cl.Cl
59548-39-9
C7H12Cl2N2O
211.086
MFCD00052003
SXCHMHOBHJOXGC-UHFFFAOYSA-N
4-methoxybenzene-1,2-diamine dihydrochloride, 4-methoxy-o-phenylenediamine dihydrochloride, 3,4-diaminoanisole dihydrochloride, 1,2-benzenediamine, 4-methoxy-, dihydrochloride, 3,4-diaminoanisole 2hcl, 4-methoxy-phenylenediamine dihydrochloride, 4-methoxy-1,2-benzenediamine dihydrochloride, 4-methoxy-benzene-1,2-diamine dihydrochloride, 4-methoxy-1,2-phenylenediamine dihydrochloride, 4-methoxy-l,2-phenylene diamine dihydrochloride
3085161
4-methoxybenzene-1,2-diamine;dihydrochloride
COC1=CC(=C(C=C1)N)N.Cl.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as derivatizing reagent by High-performance liquid chromatography (HPLC).

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents. Hygroscopic.
Melting Point ∼219°C (decomposition)
Beilstein 3699636
Quantity 25g
Solubility Information Soluble in water.
Sensitivity Hygroscopic
Formula Weight 211.09
Percent Purity ≥97%
Chemical Name or Material 4-Methoxy-o-phenylenediamine dihydrochloride

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 261-801-0

TSCA : No

Recommended Storage : Ambient temperatures

RUO – Research Use Only