Alfa Aesar™ 4-Methoxy-o-phenylenediamine dihydrochloride, 97+%

Manufacturer: Alfa Aesar™ A1236303

Quantity 1g

 View more versions of this product

Catalog No. AAA1236303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 59548-39-9
    Molecular Formula C7H10N2O·2HCl
    Formula Weight 211.09
    MDL Number MFCD00052003
    Beilstein 3699636
    Synonym 3,4-Diaminoanisole dihydrochloride
    Chemical Name or Material 4-Methoxy-o-phenylenediamine dihydrochloride

    Specifications

    Formula Weight 211.09
    CAS 59548-39-9
    Synonym 3,4-Diaminoanisole dihydrochloride
    Quantity 1g
    Health Hazard 1 Warning
    Molecular Formula C7H10N2O·2HCl
    MDL Number MFCD00052003
    Melting Point ≈219°C (dec.)
    Health Hazard 2 GHS H Statement
    H302-H312-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Beilstein 3699636
    Chemical Name or Material 4-Methoxy-o-phenylenediamine dihydrochloride
    Assay ≥97%

    Description

    Applications
    4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as derivatizing reagent by High-performance liquid chromatography (HPLC).

    Solubility
    Soluble in water.

    Notes
    Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents. Hygroscopic.

    GHS H Statement
    H302-H312-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit