Alfa Aesar™ 4'-Methoxyacetanilide, 99+%

Manufacturer: Alfa Aesar™ 3644618

Quantity 50g

 View more versions of this product

Catalog No. AA3644618

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 51-66-1
    Formula Weight 165.19
    MDL Number MFCD00014963
    Merck Index 14,51
    Synonym p-Acetanisidine; N-(4-Methoxyphenyl)acetamide
    Chemical Name or Material 4'-Methoxyacetanilide

    Specifications

    Formula Weight 165.19
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline
    Synonym p-Acetanisidine; N-(4-Methoxyphenyl)acetamide
    Health Hazard 1 Danger
    Melting Point 130° to 132°C
    Quantity 50g
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    TSCA TSCA
    MDL Number MFCD00014963
    CAS 51-66-1
    Merck Index 14,51
    Solubility Information Soluble in alcohol, acetone, chloroform; Slightly soluble in water
    Chemical Name or Material 4'-Methoxyacetanilide
    Assay ≥99%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit