Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxybenzamide 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3424-93-9
C8H9NO2
151.16
MFCD00007995
GUCPYIYFQVTFSI-UHFFFAOYSA-N
p-anisamide,p-methoxybenzamide,benzamide, 4-methoxy,4-methoxy benzamide,p-methyoxybenzamide,unii-z85fx3977e,4-methoxy-benzamide,4-anisamide,acmc-1cjjo,4-methoxylbenzamide
76959
4-methoxybenzamide
COC1=CC=C(C=C1)C(=O)N
3424-93-9
C8H9NO2
151.16
MFCD00007995
GUCPYIYFQVTFSI-UHFFFAOYSA-N
p-anisamide,p-methoxybenzamide,benzamide, 4-methoxy,4-methoxy benzamide,p-methyoxybenzamide,unii-z85fx3977e,4-methoxy-benzamide,4-anisamide,acmc-1cjjo,4-methoxylbenzamide
76959
4-methoxybenzamide
COC1=CC=C(C=C1)C(=O)N
Infrared Spectrum Authentic
Melting Point 164.0°C to 167.0°C
Color White
Quantity 25g
Packaging Glass bottle
Boiling Point 295.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3OC6H4CONH2
Beilstein 10, II, 100
Solubility Information Solubility in water: soluble
Formula Weight 151.16
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Methoxybenzamide, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 222-319-6

RTECSNumber : CV5466666