Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methoxybenzamide, 98%

Click to view available options
Quantity:
10g
50g
3424-93-9
C8H9NO2
151.165
MFCD00007995
GUCPYIYFQVTFSI-UHFFFAOYSA-N
p-anisamide,p-methoxybenzamide,benzamide, 4-methoxy,4-methoxy benzamide,p-methyoxybenzamide,unii-z85fx3977e,4-methoxy-benzamide,4-anisamide,acmc-1cjjo,4-methoxylbenzamide
76959
4-methoxybenzamide
COC1=CC=C(C=C1)C(=O)N
3424-93-9
C8H9NO2
151.165
MFCD00007995
GUCPYIYFQVTFSI-UHFFFAOYSA-N
p-anisamide,p-methoxybenzamide,benzamide, 4-methoxy,4-methoxy benzamide,p-methyoxybenzamide,unii-z85fx3977e,4-methoxy-benzamide,4-anisamide,acmc-1cjjo,4-methoxylbenzamide
76959
4-methoxybenzamide
COC1=CC=C(C=C1)C(=O)N
Melting Point 164°C to 167°C
Boiling Point 295°C
Quantity 10g
Beilstein 1862847
Formula Weight 151.17
Percent Purity 98%
Chemical Name or Material 4-Methoxybenzamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CU5466666